Assay Details
| Gene Symbol: LOC105376554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135109.1 | 2 | 2752 | 87 | ||
| GenBank mRNA | AK127718.1 | - | 2 | 2753 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dickkopf WNT signaling pathway inhibitor 3 | DKK3 |
REIC, RIG |
-49.2kb | Anti-sense | Chr. 11 - 11962996 - 12009370 on NCBI Build GRCh38 |
| Downstream |
microtubule associated monooxygenase, calponin and LIM domain containing 2 |
MICAL2 |
MICAL-2, MICAL2PV1, MICAL2PV2 |
48.8kb | Sense | Chr. 11 - 12110576 - 12281017 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105376554
Genomic Map