Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929696 | LOC101929696 | -439.0 | Anti-sense | Chr. 5 - 139741347 - 139746223 on NCBI Build GRCh38 | |
| Downstream | neuregulin 2 | NRG2 |
DON1, HRG2, NTAK |
96.8kb | Anti-sense | Chr. 5 - 139846779 - 140043675 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map