Assay Details
| Gene Symbol: THEGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL833514.1 | - | 1 | 554 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADP ribosylation factor like GTPase 9 | ARL9 | -98.8kb | Sense | Chr. 4 - 56505209 - 56525174 on NCBI Build GRCh38 | |
| Downstream | HOP homeobox | HOPX |
CAMEO, HOD, HOP, LAGY, NECC1, OB1, SMAP31, TOTO |
22.3kb | Anti-sense | Chr. 4 - 56647988 - 56681866 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

THEGL
Genomic Map