Assay Details
| Gene Symbol: LOC101930370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134241.1 | 4 | 2148 | 99 | ||
| GenBank mRNA | BC043556.1 | - | 4 | 1651 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | polypeptide N-acetylgalactosaminyltransferase-like 6 | GALNTL6 |
GALNACT20, GALNT17, GalNAc-T6L |
-103.8kb | Anti-sense | Chr. 4 - 171812690 - 173040408 on NCBI Build GRCh38 |
| Downstream | polypeptide N-acetylgalactosaminyltransferase 7 | GALNT7 |
GALNAC-T7, GalNAcT7 |
23.2kb | Anti-sense | Chr. 4 - 173168753 - 173323967 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101930370
Genomic Map