Assay Details
Gene Symbol: LOC101928004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC050584.1 | - | 1 | 3348 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | lymphocyte antigen 86 | LY86 |
MD-1, MD1, MMD-1, dJ80N2.1 |
-42.6kb | Anti-sense | Chr. 6 - 6588639 - 6654983 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC101928047 | LOC101928047 | 9.8kb | Anti-sense | Chr. 6 - 6708556 - 6730254 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
