Assay Details
| Gene Symbol: LINC00384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX648491.1 | - | 1 | 6248 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 385 | LINC00385 | Anti-sense | Chr. 13 - 30152145 - 30170093 on NCBI Build GRCh38 | ||
| Downstream | katanin catalytic subunit A1 like 1 | KATNAL1 | 43.0kb | Sense | Chr. 13 - 30202626 - 30307500 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LINC00384
Genomic Map