Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929595 | LOC101929595 | -5.2kb | Sense | Chr. 4 - 107863479 - 107932119 on NCBI Build GRCh38 | |
| Downstream | hydroxyacyl-CoA dehydrogenase | HADH |
HAD, HADH1, HADHSC, HCDH, HHF4, MSCHAD, SCHAD |
50.6kb | Anti-sense | Chr. 4 - 107989714 - 108035175 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map