Assay Details
| Gene Symbol: LOC101927009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135195.1 | 3 | 5147 | 93 | ||
| GenBank mRNA | CR749525.1 | - | 3 | 5150 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 16 open reading frame 72 | C16orf72 | PRO0149 | -283.3kb | Sense | Chr. 16 - 9091329 - 9119698 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1177 | LINC01177 | 33.2kb | Anti-sense | Chr. 16 - 9441294 - 9444985 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101927009
Genomic Map