Assay Details
| Gene Symbol: HTR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000871.2 | NP_000862.1 | 3 | 3243 | 85 | |
| GenBank mRNA | AF007141.1 | - | 1 | 1452 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MINOS1-NBL1 readthrough | MINOS1-NBL1 | C1orf151-NBL1 | -20.9kb | Sense | Chr. 1 - 19596977 - 19658456 on NCBI Build GRCh38 |
| Downstream | transmembrane and coiled-coil domains 4 | TMCO4 | 1.2kb | Anti-sense | Chr. 1 - 19682213 - 19800385 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

HTR6
Genomic Map