Assay Details
| Gene Symbol: SPAG11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL133652.1 | - | 1 | 2450 | 86 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DB340401.1 | - | 1 | Chr. 1 - 89756422 - 89756976 [+] on Build GRCh38 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 8 family member C | LRRC8C |
AD158, FAD158 |
-37.1kb | Anti-sense | Chr. 1 - 89615810 - 89719535 on NCBI Build GRCh38 |
| Downstream | leucine rich repeat containing 8 family member D | LRRC8D | LRRC5 | 61.8kb | Anti-sense | Chr. 1 - 89821014 - 89936432 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

SPAG11B
Genomic Map