Assay Details
| Gene Symbol: LOC101929398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC010527.2 | - | 1 | 852 | 80 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BQ015186.1 | - | 1 | Chr. 8 - 56494923 - 56495567 [+] on Build GRCh38 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929415 | LOC101929415 | Anti-sense | Chr. 8 - 56445807 - 56552067 on NCBI Build GRCh38 | ||
| Downstream | long intergenic non-protein coding RNA 968 | LINC00968 | 22.0kb | Sense | Chr. 8 - 56518318 - 56559823 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101929398
Genomic Map