Assay Details
| Gene Symbol: LOC105374325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC018945.2 | - | 1 | 3049 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102723362 | LOC102723362 | -963.6kb | Anti-sense | Chr. 2 - 22536479 - 22538288 on NCBI Build GRCh38 | |
| Downstream |
ATPase family, AAA domain containing 2B |
ATAD2B | 180.2kb | Sense | Chr. 2 - 23689710 - 23927114 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105374325
Genomic Map