Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger C2HC-type containing 1B | ZC2HC1B |
C6orf94, FAM164B, dJ468K18.5 |
-4.7kb | Anti-sense | Chr. 6 - 143864436 - 143938346 on NCBI Build GRCh38 |
| Downstream | hydatidiform mole associated and imprinted (non-protein coding) | HYMAI | NCRNA00020 | 57.4kb | Sense | Chr. 6 - 144004916 - 144008730 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map