Assay Details
| Gene Symbol: NPIPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR627265.1 | - | 1 | 7552 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | intraflagellar transport 43 | IFT43 |
C14orf179, CED3 |
-103.0kb | Sense | Chr. 14 - 75985753 - 76084073 on NCBI Build GRCh38 |
| Downstream | estrogen related receptor beta | ESRRB |
DFNB35, ERR2, ERRb, ESRL2, NR3B2 |
176.3kb | Sense | Chr. 14 - 76371347 - 76501839 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NPIPA1
Genomic Map