Assay Details
| Gene Symbol: MAGEA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005362.3 | NP_005353.1 | 1 | 24 | 97 | |
| XM_005274676.3 | XP_005274733.1 | 1 | 34 | 161 | ||
| GenBank mRNA | BC005963.1 | - | 1 | 24 | 4064879 | 97 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001505.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC005963.1,NM_005362.3,XM_005274676.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019560.hg.1 | TC0X001505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019560.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019560.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019559.hg.1 | TC0X001505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019559.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019559.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019562.hg.1 | TC0X001505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019562.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019562.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000417212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000417212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000417212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000417212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000417212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008750.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019561.hg.1 | TC0X001505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019561.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019561.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019563.hg.1 | TC0X001505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019563.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019563.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000412733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000412733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000412733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000412733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000412733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000457643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000457643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000457643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000457643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000457643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000417212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000417212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000417212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000417212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000417212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008750.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000329342.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000412733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000412733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000412733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000412733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000412733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000457643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000457643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000457643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000457643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000457643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | ENST00000616035.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_005363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011136.hg.1 | NM_175868.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159491.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159491.hg.1 | ENST00000417212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159491.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159491.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159491.hg.1 | ENST00000370278.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159491.hg.1 | ENST00000417212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159491.hg.1 | ENST00000598245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159491.hg.1 | NM_005362.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159491.hg.1 | TC0X00008750.hg.1 | Clariom D TC |

MAGEA3
Genomic Map