Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009580.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009580.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009580.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009580.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009580.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009580.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009580.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009580.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012227.hg.1 |
TC09000919.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012227.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012227.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012228.hg.1 |
TC09000919.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012228.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012228.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009580.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009580.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009580.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009580.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009580.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009580.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009580.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009580.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165624.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165624.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165624.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165624.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165624.hg.1 |
TC0900009580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900165625.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165625.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165625.hg.1 |
ENST00000380911.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165625.hg.1 |
NM_005454.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165625.hg.1 |
TC0900009580.hg.1 |
Clariom D TC |