Assay Details
| Gene Symbol: IGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111283.2 | NP_001104753.1 | 4 - 5 | 712 | 71 | |
| XM_017019262.1 | XP_016874751.1 | 4 - 5 | 522 | 71 | ||
| GenBank mRNA | U40870.1 | - | 3 - 4 | 391 | 71 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001890.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
skeletal system development
response to hypoxia
cell activation
platelet degranulation
DNA replication
regulation of translation
movement of cell or subcellular component
signal transduction
Ras protein signal transduction
muscle organ development
female pregnancy
aging
response to nutrient
memory
cell proliferation
positive regulation of cell proliferation
response to heat
glycolate metabolic process
regulation of gene expression
positive regulation of glycoprotein biosynthetic process
positive regulation of cardiac muscle hypertrophy
phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
response to activity
skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
muscle hypertrophy
myotube cell development
positive regulation of smooth muscle cell migration
proteoglycan biosynthetic process
positive regulation of cell migration
response to caffeine
activation of protein kinase B activity
response to lipopolysaccharide
cellular response to insulin stimulus
positive regulation of protein import into nucleus, translocation
negative regulation of smooth muscle cell apoptotic process
response to nicotine
bone mineralization involved in bone maturation
regulation of multicellular organism growth
positive regulation of activated T cell proliferation
positive regulation of tyrosine phosphorylation of Stat5 protein
negative regulation of apoptotic process
positive regulation of DNA binding
positive regulation of MAPK cascade
protein kinase B signaling
positive regulation of insulin-like growth factor receptor signaling pathway
cellular protein metabolic process
myoblast differentiation
positive regulation of fat cell differentiation
positive regulation of osteoblast differentiation
positive regulation of glycogen biosynthetic process
positive regulation of DNA replication
positive regulation of glycolytic process
positive regulation of mitotic nuclear division
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of glucose import
positive regulation of Ras protein signal transduction
insulin-like growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
positive regulation of fibroblast proliferation
positive regulation of smooth muscle cell proliferation
positive regulation of epithelial cell proliferation
positive regulation of protein secretion
positive regulation of peptidyl-tyrosine phosphorylation
protein stabilization
detection of mechanical stimulus involved in sensory perception
response to glucocorticoid
myoblast proliferation
regulation of calcium ion transport
negative regulation of oocyte development
positive regulation of cell growth involved in cardiac muscle cell development
ERK1 and ERK2 cascade
positive regulation of calcineurin-NFAT signaling cascade
cellular response to electrical stimulus
cellular response to estradiol stimulus
negative regulation of release of cytochrome c from mitochondria
negative regulation of neuron death
positive regulation of trophectodermal cell proliferation
negative regulation of cholangiocyte apoptotic process
positive regulation of transcription regulatory region DNA binding
negative regulation of extrinsic apoptotic signaling pathway
insulin receptor binding
insulin-like growth factor receptor binding
integrin binding
hormone activity
steroid binding
protein binding
growth factor activity
activation of MAPK activity
skeletal system development
response to hypoxia
cell activation
platelet degranulation
DNA replication
regulation of translation
movement of cell or subcellular component
signal transduction
Ras protein signal transduction
muscle organ development
female pregnancy
aging
response to nutrient
memory
cell proliferation
positive regulation of cell proliferation
response to heat
glycolate metabolic process
regulation of gene expression
positive regulation of glycoprotein biosynthetic process
positive regulation of cardiac muscle hypertrophy
phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
response to activity
skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
muscle hypertrophy
myotube cell development
positive regulation of smooth muscle cell migration
proteoglycan biosynthetic process
positive regulation of cell migration
response to caffeine
activation of protein kinase B activity
response to lipopolysaccharide
cellular response to insulin stimulus
positive regulation of protein import into nucleus, translocation
negative regulation of smooth muscle cell apoptotic process
response to nicotine
bone mineralization involved in bone maturation
regulation of multicellular organism growth
positive regulation of activated T cell proliferation
positive regulation of tyrosine phosphorylation of Stat5 protein
negative regulation of apoptotic process
positive regulation of DNA binding
positive regulation of MAPK cascade
protein kinase B signaling
positive regulation of insulin-like growth factor receptor signaling pathway
cellular protein metabolic process
myoblast differentiation
positive regulation of fat cell differentiation
positive regulation of osteoblast differentiation
positive regulation of glycogen biosynthetic process
positive regulation of DNA replication
positive regulation of glycolytic process
positive regulation of mitotic nuclear division
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of glucose import
positive regulation of Ras protein signal transduction
insulin-like growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
positive regulation of fibroblast proliferation
positive regulation of smooth muscle cell proliferation
positive regulation of epithelial cell proliferation
positive regulation of protein secretion
positive regulation of peptidyl-tyrosine phosphorylation
protein stabilization
detection of mechanical stimulus involved in sensory perception
response to glucocorticoid
myoblast proliferation
regulation of calcium ion transport
negative regulation of oocyte development
positive regulation of cell growth involved in cardiac muscle cell development
ERK1 and ERK2 cascade
positive regulation of calcineurin-NFAT signaling cascade
cellular response to electrical stimulus
cellular response to estradiol stimulus
negative regulation of release of cytochrome c from mitochondria
negative regulation of neuron death
positive regulation of trophectodermal cell proliferation
negative regulation of cholangiocyte apoptotic process
positive regulation of transcription regulatory region DNA binding
negative regulation of extrinsic apoptotic signaling pathway
insulin receptor binding
insulin-like growth factor receptor binding
integrin binding
hormone activity
steroid binding
protein binding
growth factor activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12024673.hg.1 | ENST00000337514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024673.hg.1 | ENST00000456098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024673.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024673.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024673.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024673.hg.1 | TC12001890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024672.hg.1 | ENST00000337514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024672.hg.1 | ENST00000456098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024672.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024672.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024672.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024672.hg.1 | TC12001890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024680.hg.1 | ENST00000337514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024680.hg.1 | ENST00000392904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024680.hg.1 | ENST00000424202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024680.hg.1 | ENST00000456098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024680.hg.1 | ENST00000481539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024680.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024680.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024680.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024680.hg.1 | TC12001890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024675.hg.1 | ENST00000337514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024675.hg.1 | ENST00000456098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024675.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024675.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024675.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024675.hg.1 | TC12001890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024678.hg.1 | ENST00000337514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024678.hg.1 | ENST00000456098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024678.hg.1 | ENST00000481539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024678.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024678.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024678.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024678.hg.1 | TC12001890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024677.hg.1 | ENST00000337514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024677.hg.1 | ENST00000456098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024677.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024677.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024677.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024677.hg.1 | TC12001890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024679.hg.1 | ENST00000337514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024679.hg.1 | ENST00000392904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024679.hg.1 | ENST00000456098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024679.hg.1 | ENST00000481539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024679.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024679.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024679.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024679.hg.1 | TC12001890.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200190847.hg.1 | ENST00000337514.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190847.hg.1 | ENST00000456098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190847.hg.1 | ENST00000481539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190847.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190847.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190847.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190847.hg.1 | ENST00000337514.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190847.hg.1 | ENST00000456098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190847.hg.1 | ENST00000481539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190847.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190847.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190847.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190847.hg.1 | TC1200011727.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | ENST00000337514.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | ENST00000392904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | ENST00000456098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | ENST00000481539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | ENST00000337514.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | ENST00000392904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | ENST00000456098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | ENST00000481539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190848.hg.1 | TC1200011727.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200190846.hg.1 | ENST00000337514.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190846.hg.1 | ENST00000456098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190846.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190846.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190846.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190846.hg.1 | ENST00000337514.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190846.hg.1 | ENST00000456098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190846.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190846.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190846.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190846.hg.1 | TC1200011727.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | ENST00000337514.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | ENST00000392904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | ENST00000424202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | ENST00000456098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | ENST00000481539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | ENST00000337514.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | ENST00000392904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | ENST00000424202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | ENST00000456098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | ENST00000481539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190849.hg.1 | TC1200011727.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200190845.hg.1 | ENST00000337514.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190845.hg.1 | ENST00000456098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190845.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190845.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190845.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190845.hg.1 | ENST00000337514.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190845.hg.1 | ENST00000456098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190845.hg.1 | NM_000618.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190845.hg.1 | NM_001111283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190845.hg.1 | NM_001111284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190845.hg.1 | TC1200011727.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 123381 | - | - |

IGF1
Genomic Map