Assay Details
Gene Symbol: OR6P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160325.1 | NP_001153797.1 | 1 | 851 | 95 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003376.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 403927 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3259693 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2148 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2149 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 1317 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3259693 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3259693 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537151 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537176 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537182 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595969 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595929 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537177 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595954 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537175 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537148 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537120 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595953 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537181 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537171 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595914 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537150 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537163 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537132 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595912 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595960 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595918 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595916 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595938 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537184 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595947 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595959 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537129 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537153 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15135139 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537137 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537155 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537139 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15135136 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595949 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595950 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537186 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595932 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595935 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595911 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595976 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537133 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595915 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595937 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537135 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595918 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537155 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537177 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595916 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537151 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15135139 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595915 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595950 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537171 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537184 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595960 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537175 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595938 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595953 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595937 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595954 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537129 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537153 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595932 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537135 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595929 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595959 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595969 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595914 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595911 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537163 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595976 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595935 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15135136 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537139 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595949 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537120 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537137 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595912 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537133 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537176 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537148 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537150 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537186 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537182 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20595947 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537181 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537132 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537171 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537182 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537121 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537151 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537128 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15135140 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537119 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537150 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15135138 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537105 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537157 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537140 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537064 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537074 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537084 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537130 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537186 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537148 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537144 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537174 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537135 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537068 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537173 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537126 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537133 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537124 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537159 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15135136 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537114 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537146 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537109 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537132 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537137 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15135135 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537063 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537117 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537077 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537120 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537169 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537176 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537179 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537111 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537188 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537107 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537167 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537082 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537183 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537160 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537070 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537164 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00172987 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7921363 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7921362 | - | - |
Affymetrix Human Genome U219 Array | 11715335_at | - | - |
Affymetrix Human Genome U219 Array | 11715335_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0100016072.hg.1 | ENST00000334632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016072.hg.1 | NM_001160325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01052751.hg.1 | TC01003376.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01052751.hg.1 | ENST00000334632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01052751.hg.1 | NM_001160325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016072.hg.1 | ENST00000334632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016072.hg.1 | NM_001160325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100222058.hg.1 | ENST00000334632.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100222058.hg.1 | NM_001160325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100222058.hg.1 | ENST00000334632.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100222058.hg.1 | NM_001160325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100222058.hg.1 | TC0100016072.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053882 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053882 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3308088 | NM_001160325.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3308088 | NM_001160325.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308088 | NM_001160325.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308088 | NM_001160325.1 | human_transcriptome |
