Assay Details
| Gene Symbol: CRLF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004750.4 | NP_004741.1 | 7 - 8 | 1409 | 103 | |
| GenBank mRNA | AF059293.1 | - | 7 - 8 | 1333 | 103 | |
| AF073515.1 | - | 7 - 8 | 1418 | 103 | ||
| AF178684.1 | - | 7 - 8 | 1266 | 103 | ||
| AY358291.1 | - | 7 - 8 | 1345 | 103 | ||
| BC044634.1 | - | 7 - 8 | 1342 | 5926400 | 103 | |
More Information
Set Membership: |
|

CRLF1
Genomic Map