Assay Details
| Gene Symbol: HNRNPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318186.1 | NP_001305115.1 | 6 - 7 | 485 | 105 | |
| NM_001318187.1 | NP_001305116.1 | 6 - 7 | 485 | 105 | ||
| NM_001318188.1 | NP_001305117.1 | 6 - 7 | 485 | 105 | ||
| NM_002140.4 | NP_002131.2 | 6 - 7 | 646 | 105 | ||
| NM_031262.3 | NP_112552.1 | 6 - 7 | 646 | 105 | ||
| NM_031263.3 | NP_112553.1 | 6 - 7 | 485 | 105 | ||
| XM_005251960.2 | XP_005252017.1 | 6 - 7 | 464 | 105 | ||
| XM_005251963.3 | XP_005252020.1 | 6 - 7 | 498 | 105 | ||
| XM_005251965.2 | XP_005252022.1 | 6 - 7 | 456 | 105 | ||
| XM_011518616.1 | XP_011516918.1 | 6 - 7 | 498 | 105 | ||
| XM_017014668.1 | XP_016870157.1 | 6 - 7 | 464 | 105 | ||
| XM_017014669.1 | XP_016870158.1 | 6 - 7 | 456 | 105 | ||
| GenBank mRNA | AB209562.1 | - | 6 - 7 | 414 | 105 | |
| AB451263.1 | - | 4 - 5 | 259 | 105 | ||
| AB451390.1 | - | 4 - 5 | 259 | 105 | ||
| AK096385.1 | - | 6 - 7 | 343 | 105 | ||
| AK291336.1 | - | 6 - 7 | 468 | 105 | ||
| AK300744.1 | - | 6 - 7 | 428 | 105 | ||
| AK307962.1 | - | 6 - 7 | 376 | 105 | ||
| AY911506.1 | - | 5 - 6 | 322 | 105 | ||
| BC000355.2 | - | 5 - 6 | 381 | 2964383 | 105 | |
| BC014980.1 | - | 6 - 7 | 452 | 4906241 | 105 | |
| BC025321.1 | - | 6 - 7 | 404 | 105 | ||
| CR456771.1 | - | 4 - 5 | 259 | 105 | ||
| DA394419.1 | - | 6 - 7 | 483 | 105 | ||
| DC318847.1 | - | 6 - 7 | 485 | 105 | ||
| S74678.1 | - | 5 - 6 | 366 | 105 | ||
| X72727.1 | - | 6 - 7 | 468 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK096385.1 transcripts.
Set Membership: |
|

HNRNPK
Genomic Map