Assay Details
| Gene Symbol: APOBEC3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181773.4 | NP_861438.3 | 4 - 5 | 670 | 107 | |
| GenBank mRNA | BQ052182.1 | - | 4 - 5 | 609 | 107 | |
More Information
Set Membership: |
|

APOBEC3H
Genomic Map