Assay Details
| Gene Symbol: NUFIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020772.2 | NP_065823.1 | 2 - 3 | 2091 | 78 | |
| XM_017024896.1 | XP_016880385.1 | 2 - 3 | 3389 | 78 | ||
| GenBank mRNA | AB037742.1 | - | 2 - 3 | 2061 | 78 | |
| AJ493465.1 | - | 2 - 3 | 2002 | 78 | ||
| AK293075.1 | - | 2 - 3 | 2091 | 78 | ||
| AY232289.1 | - | 2 - 3 | 2070 | 78 | ||
| BC108307.1 | - | 2 - 3 | 2048 | 6502841 | 78 | |
| BC129990.1 | - | 2 - 3 | 2006 | 40128910 | 78 | |
More Information
Set Membership: |
|

NUFIP2
Genomic Map