| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000579665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000579665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000579665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000579665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
ENST00000579665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000225388.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000579665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000579665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000579665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000579665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
ENST00000579665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010239.hg.1 |
NM_020772.2 |
Refseq |