Assay Details
| Gene Symbol: DLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317172.1 | NP_001304101.1 | 3 - 4 | 467 | 122 | |
| NM_003836.6 | NP_003827.3 | 3 - 4 | 467 | 122 | ||
| GenBank mRNA | AK289384.1 | - | 3 - 4 | 437 | 122 | |
| AK297918.1 | - | 2 - 3 | 363 | 122 | ||
| AK313258.1 | - | 3 - 4 | 437 | 122 | ||
| BC007741.2 | - | 3 - 4 | 416 | 4128269 | 122 | |
| BC013197.1 | - | 3 - 4 | 383 | 4347187 | 122 | |
| BC014015.2 | - | 3 - 4 | 412 | 4130556 | 122 | |
| DQ017081.1 | - | 3 - 4 | 263 | 122 | ||
| DQ126100.1 | - | 3 - 4 | 263 | 122 | ||
| DQ138965.1 | - | 3 - 4 | 263 | 122 | ||
| U15979.1 | - | 3 - 4 | 436 | 122 | ||
| U15981.1 | - | 3 - 4 | 436 | 122 | ||
| X17544.1 | - | 3 - 4 | 427 | 121 | ||
| Z12172.1 | - | 3 - 4 | 263 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the X17544.1 transcripts.
Set Membership: |
|

DLK1
Genomic Map