Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022732.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022732.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022732.hg.1 |
TC10001715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022735.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022735.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022735.hg.1 |
TC10001715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022733.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022733.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022733.hg.1 |
TC10001715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022731.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022731.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022731.hg.1 |
TC10001715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022736.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022736.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022736.hg.1 |
TC10001715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012086.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022734.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022734.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022734.hg.1 |
TC10001715.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000182653.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182653.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182653.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182653.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182653.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182653.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182653.hg.1 |
TC1000012086.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000182656.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182656.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182656.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182656.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182656.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182656.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182656.hg.1 |
TC1000012086.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000182657.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182657.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182657.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182657.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182657.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182657.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182657.hg.1 |
TC1000012086.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012086.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182655.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182655.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182655.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182655.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182655.hg.1 |
ENST00000432000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182655.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182655.hg.1 |
TC1000012086.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000182654.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182654.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182654.hg.1 |
ENST00000329446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182654.hg.1 |
NM_001010912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182654.hg.1 |
TC1000012086.hg.1 |
Clariom D TC |