Assay Details
| Gene Symbol: PABPC1L2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012977.2 | NP_001012995.1 | 1 | 2042 | 90 | |
| GenBank mRNA | AY730282.1 | - | 1 | 441 | 90 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504632 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504624 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504622 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788088 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504633 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17358017 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504627 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504629 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788101 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17358019 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788100 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32504625 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788087 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788097 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788091 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 168992 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3306232 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0950 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0949 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 7395 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P21494936 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P21494936 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3306232 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3306232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788101 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788100 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32504633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788100 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788101 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P21494936 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11788100 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P21494936 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 87018 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 73978 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21494936 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17358019 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788089 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788098 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788101 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17358017 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788095 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788092 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11788087 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P21494936 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P26998326 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001012977_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8168394 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8173598 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8173597 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8173599 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8173596 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8168393 | - | - |
| Affymetrix Human Genome U219 Array | 11737673_at | - | - |
| Affymetrix Human Genome U219 Array | 11737673_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1557203_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1557203_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040068 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040068 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 178345 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3235888 | NM_001012977.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3235888 | AY730282.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1745409 | NM_001012977.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1745409 | NM_001012977.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1745409 | NM_001012977.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3235888 | NM_001012977.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3235888 | AY730282.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3235888 | NM_001012977.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3235888 | AY730282.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1745409 | NM_001012977.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1745409 | NM_001012977.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3235888 | NM_001012977.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3235888 | AY730282.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_809-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1745409 | NM_001012977.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1745409 | NM_001012977.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 6400670 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_809 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1745409 | NM_001012977.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1745409 | NM_001012977.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1745409 | NM_001012977.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1745409 | NM_001012977.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1745409 | NM_001012977.2 | human_transcriptome |

PABPC1L2A
Genomic Map