Assay Details
| Gene Symbol: RPL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034996.2 | NP_001030168.1 | 1 - 2 | 135 | 60 | |
| GenBank mRNA | BC029036.1 | - | 1 - 2 | 123 | 5183728 | 60 |
| CD298951.1 | - | 1 - 2 | 138 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000209.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC029036.1,NM_001034996.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
ribosomal large subunit biogenesis
cell-cell adhesion
RNA binding
structural constituent of ribosome
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
ribosomal large subunit biogenesis
cell-cell adhesion
RNA binding
structural constituent of ribosome
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011972.hg.1 | ENST00000476715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004146.hg.1 | TC03000209.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004146.hg.1 | ENST00000396203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004146.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004146.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000461368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000461368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000465280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000465280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000465280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000465280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000465325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000465325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000465325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008507.hg.1 | ENST00000470084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011367.hg.1 | ENST00000424588.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | ENST00000465280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | ENST00000465325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | ENST00000465280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | ENST00000465325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203722.hg.1 | TC0300013809.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000461368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000461368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000465280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000465280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000465280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000465280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000465325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000465325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000465325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000479563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | ENST00000481798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013809.hg.1 | NM_003973.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011972.hg.1 | ENST00000476715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203716.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203716.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203716.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203716.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203716.hg.1 | ENST00000396203.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203716.hg.1 | ENST00000416518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203716.hg.1 | ENST00000435633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203716.hg.1 | NM_001034996.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203716.hg.1 | TC0300013809.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011367.hg.1 | ENST00000424588.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008507.hg.1 | ENST00000470084.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 148415 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 203700 | - | - |

RPL14
Genomic Map