Assay Details
| Gene Symbol: FER1L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039112.2 | NP_001034201.2 | 28 - 29 | 3925 | 95 | |
| XM_006716618.3 | XP_006716681.1 | 28 - 29 | 3852 | 95 | ||
| XM_011517231.2 | XP_011515533.1 | 29 - 30 | 3891 | 95 | ||
| XM_011517232.2 | XP_011515534.1 | 29 - 30 | 4032 | 95 | ||
| XM_011517233.2 | XP_011515535.1 | 28 - 29 | 3993 | 95 | ||
| XM_011517235.2 | XP_011515537.1 | 29 - 30 | 3892 | 95 | ||
| XM_011517236.2 | XP_011515538.1 | 9 - 10 | 1620 | 95 | ||
| GenBank mRNA | AB196633.1 | - | 28 - 29 | 3925 | 95 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000721.hg.1
Set Membership: |
|

FER1L6
Genomic Map