Assay Details
| Gene Symbol: FER1L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039112.2 | NP_001034201.2 | 35 - 36 | 4901 | 74 | |
| XM_006716618.3 | XP_006716681.1 | 35 - 36 | 4828 | 74 | ||
| XM_011517231.2 | XP_011515533.1 | 36 - 37 | 4867 | 74 | ||
| XM_011517232.2 | XP_011515534.1 | 36 - 37 | 5008 | 74 | ||
| XM_011517233.2 | XP_011515535.1 | 35 - 36 | 4969 | 74 | ||
| XM_011517236.2 | XP_011515538.1 | 16 - 17 | 2596 | 74 | ||
| GenBank mRNA | AB196633.1 | - | 35 - 36 | 4901 | 74 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000721.hg.1
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |

FER1L6
Genomic Map