Assay Details
| Gene Symbol: HSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040667.2 | NP_001035757.1 | 8 - 9 | 1589 | 98 | |
| NM_001538.3 | NP_001529.2 | 8 - 9 | 1589 | 98 | ||
| GenBank mRNA | AB029348.1 | - | 6 - 7 | 631 | 98 | |
| AK225152.1 | - | 6 - 7 | 646 | 98 | ||
| AK293631.1 | - | 4 - 5 | 1092 | 98 | ||
| AK296210.1 | - | 6 - 7 | 646 | 98 | ||
| BC130383.1 | - | 6 - 7 | 692 | 98 | ||
| D87673.1 | - | 6 - 7 | 631 | 98 | ||
More Information
Set Membership: |
|

HSF4
Genomic Map