| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008445.hg.1 |
TC0X000645.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008445.hg.1 |
ENST00000257013.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008445.hg.1 |
ENST00000464369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008445.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
ENST00000464369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
ENST00000464369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
ENST00000464369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
ENST00000495563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
ENST00000495563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
ENST00000495563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008443.hg.1 |
TC0X000645.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008443.hg.1 |
ENST00000257013.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008443.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008444.hg.1 |
TC0X000645.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008444.hg.1 |
ENST00000257013.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008444.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008442.hg.1 |
TC0X000645.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008442.hg.1 |
ENST00000257013.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008442.hg.1 |
ENST00000464369.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008442.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008439.hg.1 |
TC0X000645.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008439.hg.1 |
ENST00000257013.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008439.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157713.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157713.hg.1 |
ENST00000464369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157713.hg.1 |
ENST00000495563.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157713.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157713.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157713.hg.1 |
ENST00000464369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157713.hg.1 |
ENST00000495563.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157713.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157713.hg.1 |
TC0X00008495.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00157709.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157709.hg.1 |
ENST00000464369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157709.hg.1 |
ENST00000495563.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157709.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157709.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157709.hg.1 |
ENST00000464369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157709.hg.1 |
ENST00000495563.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157709.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157709.hg.1 |
TC0X00008495.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
ENST00000464369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
ENST00000464369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
ENST00000464369.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
ENST00000495563.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
ENST00000495563.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
ENST00000495563.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008495.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157711.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157711.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157711.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157711.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157711.hg.1 |
TC0X00008495.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00157705.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157705.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157705.hg.1 |
TC0X00008495.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00157710.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157710.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157710.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157710.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157710.hg.1 |
TC0X00008495.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00157712.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157712.hg.1 |
ENST00000495563.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157712.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157712.hg.1 |
ENST00000257013.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157712.hg.1 |
ENST00000495563.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157712.hg.1 |
NM_001078171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157712.hg.1 |
TC0X00008495.hg.1 |
Clariom D TC |