Assay Details
| Gene Symbol: C1GALT1C1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101330.2 | NP_001094800.1 | 1 | 749 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138127 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15237990 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138121 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138146 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138133 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218333 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218335 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218354 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218365 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218351 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218336 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138150 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138124 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218320 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218360 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138149 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218326 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138136 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138152 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218330 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218362 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138112 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138140 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218332 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218319 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138145 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218352 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138114 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138117 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218343 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218358 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138134 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218340 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138155 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01138139 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21218323 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3398513 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1650 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1651 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 62851 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3398513 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3398513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218336 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138107 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218358 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218351 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218340 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138135 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218320 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218339 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218333 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138112 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218353 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138149 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218360 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218352 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218360 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138149 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138135 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218336 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218340 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138107 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138112 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218333 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218358 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218320 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218353 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218351 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01138139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218339 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21218352 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138135 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138132 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138144 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138107 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138124 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138154 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138109 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138117 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138141 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138151 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138139 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138126 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138148 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138150 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138153 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138119 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138121 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138123 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138128 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138130 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138134 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138155 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138114 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138112 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01138138 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001101330_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8051865 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8051864 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 241710_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11758495_PM_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11758495_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11734540_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11734540_at | - | - |
| Affymetrix Human Clariom D Assay | PSR0200190662.hg.1 | NM_001101330.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200190662.hg.1 | NM_001101330.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200190662.hg.1 | TC0200012412.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 241710_at | - | - |
| Human Genome U133 Plus 2.0 Array | 241710_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241710_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241710_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241710_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241710_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241710_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241710_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241710_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241710_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241710_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241710_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241710_at5 | - | - |
| Human Genome U95 Set | 82302_at | - | - |
| Human Genome U95 Set | 82302_PM_at | - | - |
| Human Genome U133 Set | 241710_at | - | - |
| Human Genome U133 Set | 241710_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 113844 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3246294 | NM_001101330.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3246294 | NM_001101330.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3246294 | NM_001101330.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3246294 | NM_001101330.1 | human_transcriptome |

C1GALT1C1L
Genomic Map