Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000919.hg.1 |
ENST00000391428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000919.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000919.hg.1 |
TC17000074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006703.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006703.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006703.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006703.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006703.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006703.hg.1 |
ENST00000624159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006703.hg.1 |
ENST00000625754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006703.hg.1 |
ENST00000625754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006703.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006703.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006703.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006703.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000921.hg.1 |
ENST00000391428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000921.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000921.hg.1 |
TC17000074.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700146198.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146198.hg.1 |
ENST00000624159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146198.hg.1 |
ENST00000625754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146198.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146198.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146198.hg.1 |
ENST00000624159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146198.hg.1 |
ENST00000625754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146198.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146198.hg.1 |
TC1700006703.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700146199.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146199.hg.1 |
ENST00000625754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146199.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146199.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146199.hg.1 |
ENST00000625754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146199.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146199.hg.1 |
TC1700006703.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700006703.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006703.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006703.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006703.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006703.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006703.hg.1 |
ENST00000624159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006703.hg.1 |
ENST00000625754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006703.hg.1 |
ENST00000625754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006703.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006703.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006703.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006703.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146197.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146197.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146197.hg.1 |
ENST00000542475.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146197.hg.1 |
NM_001105520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146197.hg.1 |
TC1700006703.hg.1 |
Clariom D TC |