Assay Details
| Gene Symbol: ACYP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107.4 | NP_001098.1 | 1 - 2 | 103 | 96 | |
| NR_126393.1 | 1 - 2 | 103 | 96 | |||
| GenBank mRNA | AK312101.1 | - | 1 - 2 | 103 | 96 | |
| BC035568.1 | - | 1 - 2 | 88 | 5392248 | 96 | |
| BG614847.1 | - | 1 - 2 | 70 | 96 | ||
| DB071115.1 | - | 1 - 2 | 103 | 96 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001316.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK312101.1,BC035568.1,NM_001107.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14015050.hg.1 | ENST00000238618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015050.hg.1 | ENST00000357971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015050.hg.1 | ENST00000553302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015050.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015050.hg.1 | ENST00000555694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015050.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015050.hg.1 | TC14001316.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000238618.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000238618.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000238618.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000238618.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000553302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000553302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000553302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000555463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000555463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000555463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000555463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000555694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000555694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000555694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | ENST00000555694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NM_001302616.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NM_001302616.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NM_001302616.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NM_001302616.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NM_001302617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NM_001302617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NM_001302617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NM_001302617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NR_126394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NR_126394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NR_126394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NR_126395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NR_126395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009710.hg.1 | NR_126395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015048.hg.1 | ENST00000238618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015048.hg.1 | ENST00000357971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015048.hg.1 | ENST00000553302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015048.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015048.hg.1 | ENST00000555463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015048.hg.1 | ENST00000555694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015048.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015048.hg.1 | TC14001316.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | ENST00000238618.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | ENST00000553302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | ENST00000555694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | NM_001302616.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | NR_126395.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | ENST00000238618.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | ENST00000553302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | ENST00000555694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | NM_001302616.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | NR_126395.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171694.hg.1 | TC1400009710.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | ENST00000238618.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | ENST00000553302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | ENST00000555463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | ENST00000555694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | NM_001302616.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | NM_001302617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | ENST00000238618.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | ENST00000553302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | ENST00000555463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | ENST00000555694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | NM_001302616.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | NM_001302617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400171691.hg.1 | TC1400009710.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000238618.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000238618.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000238618.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000238618.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000357971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000553302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000553302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000553302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000555135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000555463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000555463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000555463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000555463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000555694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000555694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000555694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | ENST00000555694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NM_001107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NM_001302616.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NM_001302616.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NM_001302616.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NM_001302616.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NM_001302617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NM_001302617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NM_001302617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NM_001302617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NR_126393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NR_126394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NR_126394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NR_126394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NR_126395.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NR_126395.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009710.hg.1 | NR_126395.1 | Refseq |

ACYP1
Genomic Map