Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004570.hg.1 |
TC07000303.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004570.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004570.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004571.hg.1 |
TC07000303.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004571.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004571.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007467.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007467.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007467.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007467.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007467.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007467.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004572.hg.1 |
TC07000303.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004572.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004572.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007467.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007467.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007467.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007467.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007467.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007467.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151927.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151927.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151927.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151927.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151927.hg.1 |
TC0700007467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700151928.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151928.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151928.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151928.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151928.hg.1 |
TC0700007467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700151929.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151929.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151929.hg.1 |
ENST00000408988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151929.hg.1 |
NM_001123065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151929.hg.1 |
TC0700007467.hg.1 |
Clariom D TC |