Assay Details
| Gene Symbol: THTPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126339.3 | NP_001119811.1 | 2 - 3 | 923 | 89 | |
| NM_024328.5 | NP_077304.1 | 1 - 2 | 1322 | 89 | ||
| GenBank mRNA | AF432862.1 | - | 1 - 2 | 591 | 89 | |
| AK057691.1 | - | 1 - 2 | 1029 | 89 | ||
| AK098604.1 | - | 2 - 3 | 712 | 89 | ||
| BC002984.1 | - | 1 - 2 | 1016 | 3535451 | 89 | |
| BI522644.1 | - | 2 - 3 | 727 | 89 | ||
| BX161435.1 | - | 1 - 2 | 1282 | 89 | ||
More Information
Set Membership: |
|

THTPA
Genomic Map