Assay Details
| Gene Symbol: SLC36A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181776.2 | NP_861441.2 | 8 - 9 | 1137 | 88 | |
| XM_005268377.3 | XP_005268434.1 | 8 - 9 | 1132 | 88 | ||
| XM_006714756.3 | XP_006714819.1 | 7 - 8 | 1033 | 88 | ||
| XM_017009084.1 | XP_016864573.1 | 7 - 8 | 1045 | 88 | ||
| GenBank mRNA | AK309575.1 | - | 8 - 9 | 1137 | 88 | |
| AY162214.1 | - | 8 - 9 | 1105 | 88 | ||
| BC101100.1 | - | 8 - 9 | 1087 | 88 | ||
| BC101101.1 | - | 8 - 9 | 1075 | 40010274 | 88 | |
| BC101102.1 | - | 7 - 8 | 990 | 40010277 | 88 | |
| BC101103.2 | - | 8 - 9 | 1075 | 40010278 | 88 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001951.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ion transport
amino acid transport
L-alanine transport
glycine transport
proline transport
proline transmembrane transport
hydrogen ion transmembrane transport
hydrogen:amino acid symporter activity
hydrogen ion transmembrane transporter activity
amino acid transmembrane transporter activity
L-alanine transmembrane transporter activity
glycine transmembrane transporter activity
L-proline transmembrane transporter activity
ion transport
amino acid transport
L-alanine transport
glycine transport
proline transport
proline transmembrane transport
hydrogen ion transmembrane transport
hydrogen:amino acid symporter activity
hydrogen ion transmembrane transporter activity
amino acid transmembrane transporter activity
L-alanine transmembrane transporter activity
glycine transmembrane transporter activity
L-proline transmembrane transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05027180.hg.1 | TC05001951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027180.hg.1 | ENST00000335244.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027180.hg.1 | ENST00000450886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027180.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027180.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000523044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000523044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | ENST00000523044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000522829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000523044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000523044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | ENST00000523044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012506.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187288.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187288.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187288.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187288.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187288.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187288.hg.1 | ENST00000335244.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187288.hg.1 | ENST00000518280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187288.hg.1 | ENST00000518617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187288.hg.1 | ENST00000521967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187288.hg.1 | NM_181776.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187288.hg.1 | TC0500012506.hg.1 | Clariom D TC |

SLC36A2
Genomic Map