Assay Details
| Gene Symbol: SCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169110.1 | NP_001162581.1 | 1 - 2 | 145 | 64 | |
| GenBank mRNA | BG392981.1 | - | 1 - 2 | 146 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001169110.1 transcripts.
Set Membership: |
|

SCO2
Genomic Map