Assay Details
| Gene Symbol: TNFRSF10D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003840.4 | NP_003831.2 | 4 - 5 | 580 | 102 | |
| GenBank mRNA | AF021232.1 | - | 4 - 5 | 563 | 102 | |
| AF021233.1 | - | 4 - 5 | 564 | 102 | ||
| AF023849.1 | - | 4 - 5 | 485 | 102 | ||
| AF029761.1 | - | 4 - 5 | 567 | 102 | ||
| AK313528.1 | - | 4 - 5 | 577 | 102 | ||
| AY358285.1 | - | 4 - 5 | 568 | 102 | ||
| BC052270.1 | - | 4 - 5 | 568 | 6572696 | 102 | |
More Information
Set Membership: |
|

TNFRSF10D
Genomic Map