Assay Details
| Gene Symbol: ANP32C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012403.1 | NP_036535.1 | 1 | 249 | 120 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001702.hg.1
Set Membership: |
|

ANP32C
Genomic Map