Assay Details
| Gene Symbol: SYNM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015286.5 | NP_056101.5 | 3 - 4 | 1126 | 89 | |
| NM_145728.2 | NP_663780.2 | 3 - 4 | 1126 | 89 | ||
| XM_017022035.1 | XP_016877524.1 | 3 - 4 | 1137 | 89 | ||
| GenBank mRNA | AB002351.2 | - | 3 - 4 | 1164 | 89 | |
| AF359284.1 | - | 3 - 4 | 1117 | 89 | ||
| AJ310521.1 | - | 3 - 4 | 1120 | 89 | ||
| AJ310522.1 | - | 3 - 4 | 1120 | 89 | ||
| AL833262.1 | - | 3 - 4 | 617 | 89 | ||
| BC151243.1 | - | 3 - 4 | 1171 | 8860516 | 89 | |
More Information
Set Membership: |
|

SYNM
Genomic Map