Assay Details
| Gene Symbol: TOMM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134485.1 | NP_001127957.1 | 1 - 2 | 331 | 98 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09002890.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014426.hg.1 | TC09002890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014426.hg.1 | ENST00000377773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014426.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014426.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014426.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014426.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900185945.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900185945.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900185945.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900185945.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900185945.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900185945.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900185945.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900185945.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900185945.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900185945.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900185945.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900185945.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900185945.hg.1 | TC0900012228.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000321301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000377773.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000401811.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000540941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | ENST00000544379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001001790.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012228.hg.1 | NM_001134485.1 | Refseq |

TOMM5
Genomic Map