Assay Details
| Gene Symbol: JDP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135047.1 | NP_001128519.1 | 1 - 2 | 46 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000473.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001135047.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
positive regulation of histone deacetylation
negative regulation of fat cell differentiation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
protein binding
protein heterodimerization activity
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
positive regulation of histone deacetylation
negative regulation of fat cell differentiation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
protein binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000558068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000558068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000558068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000558068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000559773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000559773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000559773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | ENST00000559773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005890.hg.1 | ENST00000419727.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005890.hg.1 | ENST00000559773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005890.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005890.hg.1 | TC14000473.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005894.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005894.hg.1 | ENST00000419727.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005894.hg.1 | ENST00000435893.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005894.hg.1 | ENST00000437176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005894.hg.1 | ENST00000559773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005894.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005894.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005894.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005894.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005894.hg.1 | TC14000473.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000558068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000559773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000558068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | ENST00000559773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154503.hg.1 | TC1400007710.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000267569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000435893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000437176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000558068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000558068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000558068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000558068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000559060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000559773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000559773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000559773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | ENST00000559773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_001135049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007710.hg.1 | NM_130469.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154495.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154495.hg.1 | ENST00000559773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154495.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154495.hg.1 | ENST00000419727.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154495.hg.1 | ENST00000559773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154495.hg.1 | NM_001135047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154495.hg.1 | TC1400007710.hg.1 | Clariom D TC |

JDP2
Genomic Map