Assay Details
| Gene Symbol: TP53INP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135733.1 | NP_001129205.1 | 3 - 4 | 854 | 70 | |
| GenBank mRNA | AF409114.1 | - | 3 - 4 | 826 | 70 | |
| AK125880.1 | - | 3 - 4 | 833 | 70 | ||
| AL833652.1 | - | 2 - 3 | 434 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

TP53INP1
Genomic Map