Assay Details
| Gene Symbol: LITAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136472.1 | NP_001129944.1 | 1 - 2 | 61 | 83 | |
| NM_001136473.1 | NP_001129945.1 | 1 - 2 | 61 | 83 | ||
| GenBank mRNA | AK095955.1 | - | 1 - 2 | 61 | 83 | |
| AK130788.1 | - | 1 - 2 | 61 | 83 | ||
| AK314519.1 | - | 1 - 2 | 61 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001136472.1, NM_001136473.1 transcripts.
Set Membership: |
|

LITAF
Genomic Map