Assay Details
| Gene Symbol: NFYC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142589.1 | NP_001136061.1 | 3 - 4 | 477 | 85 | |
| GenBank mRNA | AK301385.1 | - | 3 - 4 | 361 | 85 | |
More Information
Set Membership: |
|

NFYC
Genomic Map