Assay Details
| Gene Symbol: PTPMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143984.1 | NP_001137456.1 | 1 - 2 | 561 | 102 | |
| GenBank mRNA | AF277187.1 | - | 1 - 2 | 58 | 102 | |
| AK294640.1 | - | 2 - 3 | 283 | 102 | ||
| AY333987.1 | - | 2 - 3 | 259 | 102 | ||
| BC107573.1 | - | 1 - 2 | 396 | 102 | ||
More Information
Set Membership: |
|

PTPMT1
Genomic Map