Assay Details
Gene Symbol: RFPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159545.1 | NP_001153017.1 | 1 - 2 | 28 | 52 | |
XM_011529827.2 | XP_011528129.1 | 1 - 2 | 105 | 52 | ||
XM_017028535.1 | XP_016884024.1 | 1 - 2 | 141 | 52 | ||
GenBank mRNA | BC051910.1 | - | 1 - 2 | 28 | 5729832 | 52 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC051910.1,NM_001159545.1,XM_011529827.2,XM_017028535.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000489846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000489846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000489846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000489846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000489846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000628378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000628378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000628378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000628378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000628378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | ENST00000628378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200161244.hg.1 | TC2200008505.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200161244.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200161244.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000248983.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000400237.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000489846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000489846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000489846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000489846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000489846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000626996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000628378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000628378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000628378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000628378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000628378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | ENST00000628378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001098527.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159545.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_001159546.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008505.hg.1 | NM_006605.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 128141 | - | - |
