Assay Details
| Gene Symbol: MAFF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161572.1 | NP_001155044.1 | 1 - 2 | 307 | 77 | |
| GenBank mRNA | DA654696.1 | - | 1 - 2 | 249 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000297.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001161572.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
parturition
blood coagulation
skeletal muscle cell differentiation
regulation of epidermal cell differentiation
positive regulation of transcription from RNA polymerase II promoter
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
sequence-specific DNA binding
in utero embryonic development
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
parturition
blood coagulation
skeletal muscle cell differentiation
regulation of epidermal cell differentiation
positive regulation of transcription from RNA polymerase II promoter
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22005609.hg.1 | ENST00000538320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005609.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005609.hg.1 | TC22000297.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000417948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000417948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000417948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000417948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000417948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000417948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000441709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000441709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000441709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000441709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200151702.hg.1 | TC2200007337.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200151702.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200151702.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200151702.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200151702.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200151702.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200151702.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000338483.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000407965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000417948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000417948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000417948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000417948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000417948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000417948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000426621.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000441709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000441709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000441709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000441709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000538999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | ENST00000624676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_001161574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_012323.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007337.hg.1 | NM_012323.3 | Refseq |

MAFF
Genomic Map