Assay Details
| Gene Symbol: CHAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164144.1 | NP_001157616.1 | 2 - 3 | 278 | 90 | |
| GenBank mRNA | DA289328.1 | - | 2 - 3 | 247 | 90 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000437.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001164144.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13005006.hg.1 | ENST00000361283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005006.hg.1 | ENST00000463003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005006.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005006.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005006.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005006.hg.1 | TC13000437.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005008.hg.1 | ENST00000361283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005008.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005008.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005008.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005008.hg.1 | TC13000437.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005005.hg.1 | ENST00000361283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005005.hg.1 | ENST00000463003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005005.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005005.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005005.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005005.hg.1 | TC13000437.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005007.hg.1 | ENST00000361283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005007.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005007.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005007.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005007.hg.1 | TC13000437.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | ENST00000463003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | ENST00000463003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | ENST00000463003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | ENST00000478022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | ENST00000478022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155160.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155160.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155160.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155160.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155160.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155160.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155160.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155160.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155160.hg.1 | TC1300008181.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300155159.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155159.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155159.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155159.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155159.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155159.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155159.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155159.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155159.hg.1 | TC1300008181.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300155157.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155157.hg.1 | ENST00000463003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155157.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155157.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155157.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155157.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155157.hg.1 | ENST00000463003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155157.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155157.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155157.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155157.hg.1 | TC1300008181.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | ENST00000463003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | ENST00000463003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | ENST00000463003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | ENST00000478022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | ENST00000478022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008181.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155158.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155158.hg.1 | ENST00000463003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155158.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155158.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155158.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155158.hg.1 | ENST00000361283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155158.hg.1 | ENST00000463003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155158.hg.1 | NM_001164144.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155158.hg.1 | NM_001164145.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155158.hg.1 | NM_032436.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155158.hg.1 | TC1300008181.hg.1 | Clariom D TC |

CHAMP1
Genomic Map